Assay Details
| Gene Symbol: LOC100129072 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK123881.1 | - | 3 | 1553 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 85C | CCDC85C | Anti-sense | Chr. 14 - 99511266 - 99604390 on NCBI Build GRCh38 | ||
| Downstream | HHIP like 1 | HHIPL1 |
KIAA1822, UNQ9245 |
81.9kb | Sense | Chr. 14 - 99604476 - 99680569 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100129072
Genomic Map