Assay Details
| Gene Symbol: C14orf144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC007749.1 | - | 1 | 49 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kinesin family member 26A | KIF26A | -63.3kb | Sense | Chr. 14 - 104138723 - 104180898 on NCBI Build GRCh38 | |
| Downstream | chromosome 14 open reading frame 180 | C14orf180 |
C14orf77, NRAC |
335.2kb | Sense | Chr. 14 - 104579684 - 104589847 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530466 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530419 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530475 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994407 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530426 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530444 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994464 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994374 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530429 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530421 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994458 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530462 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530485 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530478 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530468 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16738714 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530464 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16738715 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530436 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994421 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530459 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530448 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530433 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530420 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530457 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994472 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530495 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994380 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994450 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530477 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530498 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530470 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530460 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994405 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530435 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530452 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530494 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530430 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16738709 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530490 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994447 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994384 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530496 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530461 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16738710 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530431 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994439 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994443 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994445 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530483 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530427 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530428 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530486 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530450 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530455 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994471 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530425 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530447 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994470 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994422 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530458 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530418 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994413 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994404 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994482 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P29994475 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530497 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530443 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530471 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09530424 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3449815 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P180101 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 39635 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 23748 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 15106 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P180101 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P180101 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P180102 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P180102 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 181769 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1669545 | - | - |
| HumanHT-12 v4 Expression BeadChip | ILMN_1760222 | BC007749.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1760222 | BC007749.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1669545 | - | - |
| HumanHT-12 V4.0 expression beadchip | ILMN_1669545 | - | - |
| HumanHT-12 V4.0 expression beadchip | ILMN_1760222 | BC007749.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1669545 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1760222 | BC007749.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_30862 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_30862 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1669545 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1760222 | BC007749.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1669545 | - | - |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1760222 | BC007749.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1760222 | BC007749.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1669545 | - | - |

C14orf144
Genomic Map