Assay Details
Gene Symbol: LOC400756 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC030752.2 | - | 2 | 1055 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | dedicator of cytokinesis 7 | DOCK7 |
EIEE23, ZIR2 |
-21.3kb | Anti-sense | Chr. 1 - 62454726 - 62688368 on NCBI Build GRCh38 |
Downstream | autophagy related 4C cysteine peptidase | ATG4C |
APG4-C, APG4C, AUTL1, AUTL3 |
73.4kb | Sense | Chr. 1 - 62784106 - 62865270 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00244273 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00244286 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272540 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00244277 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272533 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272550 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272562 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272577 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272532 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00244258 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00244276 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00244271 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272522 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272518 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272536 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272520 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00244285 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272572 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272537 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272515 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272530 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272531 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00244298 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272564 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272553 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272524 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272516 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272543 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272555 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00244267 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272525 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272521 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272556 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272549 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00244272 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272554 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00244255 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00244270 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00244287 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20272570 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00244295 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00244259 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 122788 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 85200 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 56685 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 149214 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 200895 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 777229 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 589689 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 578653 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 834159 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 773375 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 40657 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 207666 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 251705 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3620087 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P887687 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 45002 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 6964 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3620087 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P887687 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P15144913 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P15144913 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P15144925 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P887687 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 72444 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 169915 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 81519 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15144925 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15144924 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P10142549 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P10143427 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P15144913 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P35152679 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P10144935 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00080441 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P887687 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15144923 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15144913 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P10143427 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P15144925 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 1556255_PM_a_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0100008562.hg.1 | ENST00000453229.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008562.hg.1 | ENST00000453229.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008562.hg.1 | ENST00000453229.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008562.hg.1 | ENST00000453229.1 | Ensembl |
Human Genome U133 Plus 2.0 Array | 1556255_a_at | - | - |
Human Genome U133 Plus 2.0 Array | 1556255_PM_a_at | - | - |
Human Genome U133 Plus 2.0 Array | 1561158_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1561158_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556255_a_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556255_a_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556255_a_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556255_a_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556255_a_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556255_a_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556255_a_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556255_a_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556255_a_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556255_a_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556255_a_at11 | - | - |
Human Genome U95 Set | 67272_at | - | - |
Human Genome U95 Set | 67272_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 709258 | - | - |
