Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT HG-U133+ PM Array Plate |
1557566_PM_at |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
1557567_PM_a_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010968.hg.1 |
TC01000676.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010968.hg.1 |
NR_038320.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010969.hg.1 |
TC01000676.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010969.hg.1 |
ENST00000499090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010969.hg.1 |
NR_038320.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010967.hg.1 |
TC01000676.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010967.hg.1 |
ENST00000499090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01010967.hg.1 |
NR_038320.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159961.hg.1 |
NR_038320.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159961.hg.1 |
NR_038320.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159961.hg.1 |
TC0100008420.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100159963.hg.1 |
NR_038320.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159963.hg.1 |
NR_038320.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159963.hg.1 |
TC0100008420.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100159959.hg.1 |
NR_038320.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159959.hg.1 |
NR_038320.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100159959.hg.1 |
TC0100008420.hg.1 |
Clariom D TC |
Human Genome U133 Plus 2.0 Array |
1557566_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1557566_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1557567_a_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
1557567_PM_a_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557567_a_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557566_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557566_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557566_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557566_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557567_a_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557566_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557567_a_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557567_a_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557566_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557566_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557567_a_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557566_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557567_a_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557567_a_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557566_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557567_a_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557566_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557567_a_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557567_a_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557567_a_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
1557566_at5 |
- |
- |