Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010003.hg.1 |
ENST00000514871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010003.hg.1 |
ENST00000514871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010003.hg.1 |
ENST00000563727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010003.hg.1 |
ENST00000563727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010003.hg.1 |
ENST00000563727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010003.hg.1 |
NR_027073.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010003.hg.1 |
NR_027073.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010003.hg.1 |
ENST00000514871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010003.hg.1 |
ENST00000514871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010003.hg.1 |
ENST00000563727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010003.hg.1 |
ENST00000563727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010003.hg.1 |
ENST00000563727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010003.hg.1 |
NR_027073.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010003.hg.1 |
NR_027073.1 |
Refseq |