Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010114.hg.1 |
ENST00000522004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010114.hg.1 |
ENST00000522004.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010658.hg.1 |
NR_040117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010658.hg.1 |
NR_040117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010658.hg.1 |
NR_040117.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010658.hg.1 |
NR_040117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010114.hg.1 |
ENST00000522004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010114.hg.1 |
ENST00000522004.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010658.hg.1 |
ENST00000399186.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010658.hg.1 |
NR_040117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010658.hg.1 |
NR_040117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010658.hg.1 |
NR_040117.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010658.hg.1 |
NR_040117.1 |
Refseq |