| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017747.hg.1 |
TC0X001323.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017747.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017747.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017747.hg.1 |
ENST00000555168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017747.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
ENST00000536487.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
ENST00000536487.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
ENST00000585790.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
ENST00000585790.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00011392.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017749.hg.1 |
TC0X001323.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017749.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017749.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017748.hg.1 |
TC0X001323.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017748.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017748.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017748.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00179009.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179009.hg.1 |
ENST00000536487.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179009.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179009.hg.1 |
ENST00000585790.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179009.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00179009.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179009.hg.1 |
ENST00000536487.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179009.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179009.hg.1 |
ENST00000585790.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179009.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00179009.hg.1 |
TC0X00011392.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
ENST00000536487.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
ENST00000536487.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
ENST00000585790.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
ENST00000585790.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00011392.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00179013.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179013.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00179013.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179013.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00179013.hg.1 |
TC0X00011392.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00179011.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179011.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179011.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00179011.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179011.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179011.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00179011.hg.1 |
TC0X00011392.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00179010.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179010.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179010.hg.1 |
ENST00000585790.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179010.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00179010.hg.1 |
ENST00000452254.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179010.hg.1 |
ENST00000554109.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179010.hg.1 |
ENST00000585790.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00179010.hg.1 |
NR_027131.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00179010.hg.1 |
TC0X00011392.hg.1 |
Clariom D TC |