Assay Details
| Gene Symbol: SLC32A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF064848.1 | - | 1 | 49 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rho GTPase activating protein 40 | ARHGAP40 |
C20orf95, dJ1100H13.4 |
-73.6kb | Sense | Chr. 20 - 38601934 - 38650652 on NCBI Build GRCh38 |
| Downstream | ARP5 actin-related protein 5 homolog | ACTR5 |
Arp5, INO80M |
23.7kb | Sense | Chr. 20 - 38748454 - 38772446 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SLC32A1
Genomic Map