Assay Details
Gene Symbol: TPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK021810.1 | - | 1 | 1043 | 106 | |
AK130640.1 | - | 1 | 746 | 106 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | NOBOX oogenesis homeobox | NOBOX |
OG-2, OG2, OG2X, POF5, TCAG_12042 |
-314.6kb | Sense | Chr. 7 - 144396805 - 144410438 on NCBI Build GRCh38 |
Downstream | contactin associated protein-like 2 | CNTNAP2 |
AUTS15, CASPR2, CDFE, NRXN4, PTHSL1 |
1.4mb | Anti-sense | Chr. 7 - 146116035 - 148420998 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
