Assay Details
Gene Symbol: DEPTOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK022403.1 | - | 1756 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | DNA replication and sister chromatid cohesion 1 | DSCC1 | DCC1 | -181.0kb | Sense | Chr. 8 - 119833941 - 119855930 on NCBI Build GRCh38 |
Downstream | collagen type XIV alpha 1 chain | COL14A1 | UND | 87.6kb | Anti-sense | Chr. 8 - 120124468 - 120373575 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
