Assay Details
Gene Symbol: TDRD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK023578.1 | - | 1446 | 113 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 434 | LINC00434 | -304.8kb | Anti-sense | Chr. 13 - 60214347 - 60268104 on NCBI Build GRCh38 | |
Downstream | long intergenic non-protein coding RNA 378 | LINC00378 | 99.4kb | Sense | Chr. 13 - 60672955 - 60695806 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
