Assay Details
Gene Symbol: ZNF391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK092633.1 | - | 1 | 1749 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
zinc finger protein 204, pseudogene |
ZNF204P |
ZNF184-Lp, ZNF204, ZNF315P, b24o18.1 |
-13.4kb | Anti-sense | Chr. 6 - 27357823 - 27375374 on NCBI Build GRCh38 |
Downstream | zinc finger protein 184 | ZNF184 | kr-ZNF3 | 59.8kb | Anti-sense | Chr. 6 - 27450742 - 27473118 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
