Array Name |
Probe IDs |
Targets |
Target Type |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
86921 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
342429 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_32_P876086 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
27049 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
29077 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P40297022 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_32_P876086 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_16_P40297022 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P40297022 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
114096 |
AK225532.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
114096 |
AK312900.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
114096 |
AL833628.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
114096 |
BC006991.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
114096 |
CR591791.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
114096 |
CR610035.1 |
genbank_mrna_human |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
114096 |
NM_001104546.1 |
refseq_mrna_human_NM |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
114096 |
NM_006413.4 |
refseq_mrna_human_NM |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
114096 |
U77665.1 |
genbank_mrna_human |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
89831 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P33604222 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P40297022 |
- |
- |
Agilent Human 1B Microarray G4111A |
A_32_P876086 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P40297022 |
- |
- |