Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007903.hg.1 |
ENST00000611039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007903.hg.1 |
ENST00000611039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007903.hg.1 |
ENST00000611039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007903.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007903.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007903.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007903.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007903.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200156156.hg.1 |
TC2200007903.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200156156.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156156.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156156.hg.1 |
ENST00000611039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156156.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200156156.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156156.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156156.hg.1 |
ENST00000611039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156156.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200156153.hg.1 |
TC2200007903.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200156153.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156153.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156153.hg.1 |
ENST00000611039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156153.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200156153.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156153.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156153.hg.1 |
ENST00000611039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156153.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200156155.hg.1 |
TC2200007903.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200156155.hg.1 |
ENST00000611039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156155.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200156155.hg.1 |
ENST00000611039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156155.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200156157.hg.1 |
TC2200007903.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200156157.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156157.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156157.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200156157.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156157.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156157.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200156151.hg.1 |
TC2200007903.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200156151.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156151.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156151.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200156151.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156151.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156151.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007903.hg.1 |
ENST00000443935.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007903.hg.1 |
ENST00000611039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007903.hg.1 |
ENST00000611039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007903.hg.1 |
ENST00000611039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007903.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007903.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007903.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007903.hg.1 |
NM_001288707.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007903.hg.1 |
NM_001288707.1 |
Refseq |