Assay Details
| Gene Symbol: XYLT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC017084.2 | - | 1 | 853 | 114 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | T87501.1 | - | 1 | Chr. 17 - 50357913 - 50358176 [-] on Build GRCh38 | 113 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TMEM92 antisense RNA 1 | TMEM92-AS1 |
TCONS_00025237, lncRNA-508851 |
-69.3kb | Anti-sense | Chr. 17 - 50281579 - 50287855 on NCBI Build GRCh38 |
| Downstream | mitochondrial ribosomal protein L27 | MRPL27 | L27mt | 9.8kb | Anti-sense | Chr. 17 - 50367867 - 50373201 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000509778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000509778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000571021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000571021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000571021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000574840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | ENST00000574840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000017003.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000376550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000507602.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000509778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000509778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000511654.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000571021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000571021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000571021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000574840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | ENST00000574840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NM_022167.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008250.hg.1 | NR_110010.1 | Refseq |

XYLT2
Genomic Map