Assay Details
| Gene Symbol: CX3CR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC017855.1 | - | 1 | 1246 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | xin actin binding repeat containing 1 | XIRP1 |
CMYA1, Xin |
-86.0kb | Sense | Chr. 3 - 39183214 - 39192621 on NCBI Build GRCh38 |
| Downstream | C-C motif chemokine receptor 8 | CCR8 |
CC-CKR-8, CCR-8, CDw198, CKRL1, CMKBR8, CMKBRL2, CY6, GPRCY6, TER1 |
49.7kb | Anti-sense | Chr. 3 - 39329706 - 39333680 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

CX3CR1
Genomic Map