Assay Details
| Gene Symbol: TMEM80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC038383.1 | - | 1 | 1049 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
DEAF1, transcription factor |
DEAF1 |
MRD24, NUDR, SPN, ZMYND5 |
Anti-sense | Chr. 11 - 644220 - 695754 on NCBI Build GRCh38 | |
| Downstream | EPS8 like 2 | EPS8L2 | EPS8R2 | 9.7kb | Sense | Chr. 11 - 705217 - 727727 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TMEM80
Genomic Map