Assay Details
Gene Symbol: CARD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC042201.1 | - | 1 | 2051 | 107 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | sushi domain containing 3 | SUSD3 | -11.1kb | Sense | Chr. 9 - 93058688 - 93085138 on NCBI Build GRCh38 | |
Downstream | ninjurin 1 | NINJ1 |
NIN1, NINJURIN |
23.1kb | Anti-sense | Chr. 9 - 93121489 - 93134304 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
