Assay Details
| Gene Symbol: SMPDL3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005266797.4 | XP_005266854.1 | 1 | 49 | 79 | |
| GenBank mRNA | BC066591.1 | - | 1 | 550 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | fatty acid binding protein 7 | FABP7 |
B-FABP, BLBP, FABPB, MRG |
-5.2kb | Sense | Chr. 6 - 122749201 - 122784077 on NCBI Build GRCh38 |
| Downstream | clavesin 2 | CLVS2 |
C6orf212, C6orf213, RLBP1L2, bA160A10.4 |
205.2kb | Sense | Chr. 6 - 122993693 - 123081282 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005266797.4 transcripts.
Set Membership: |
|

SMPDL3A
Genomic Map