Assay Details
Gene Symbol: ABCB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163941.1 | NP_001157413.1 | 13 - 14 | 1770 | 91 | |
NM_001163942.1 | NP_001157414.1 | 4 - 5 | 744 | 91 | ||
NM_001163993.2 | NP_001157465.1 | 4 - 5 | 744 | 91 | ||
NM_178559.5 | NP_848654.3 | 4 - 5 | 744 | 91 | ||
XM_011515367.2 | XP_011513669.1 | 5 - 6 | 690 | 91 | ||
GenBank mRNA | AB353947.1 | - | 13 - 14 | 1770 | 91 | |
AF399931.1 | - | 2 - 3 | 258 | 91 | ||
AY090613.1 | - | 4 - 5 | 665 | 91 | ||
AY230001.1 | - | 13 - 14 | 1663 | 91 | ||
AY234788.1 | - | 4 - 5 | 665 | 91 | ||
AY851364.1 | - | 4 - 5 | 758 | 91 | ||
AY851365.1 | - | 4 - 5 | 551 | 91 | ||
BC104894.1 | - | 4 - 5 | 494 | 8143897 | 91 | |
BC104920.1 | - | 4 - 5 | 492 | 8143923 | 91 | |
GU437216.1 | - | 1 - 2 | 198 | 91 | ||
GU437217.1 | - | 1 - 2 | 198 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
