| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027759.hg.1 |
TC01001775.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027759.hg.1 |
ENST00000366988.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027759.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027759.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027759.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027763.hg.1 |
TC01001775.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027763.hg.1 |
ENST00000366988.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027763.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027763.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027763.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000472389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000472389.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027755.hg.1 |
TC01001775.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027755.hg.1 |
ENST00000366988.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027755.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027755.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027757.hg.1 |
TC01001775.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027757.hg.1 |
ENST00000366988.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027757.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01027757.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184889.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184889.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184889.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184889.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184889.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184889.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184889.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184889.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184889.hg.1 |
TC0100011527.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100184895.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184895.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184895.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184895.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184895.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184895.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184895.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184895.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184895.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184895.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184895.hg.1 |
TC0100011527.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000472389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000472389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100011527.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184886.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184886.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184886.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184886.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184886.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184886.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184886.hg.1 |
TC0100011527.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100184891.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184891.hg.1 |
ENST00000472389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184891.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184891.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184891.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184891.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184891.hg.1 |
ENST00000366988.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184891.hg.1 |
ENST00000472389.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184891.hg.1 |
ENST00000473900.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184891.hg.1 |
ENST00000479589.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100184891.hg.1 |
NM_013349.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184891.hg.1 |
NR_026598.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100184891.hg.1 |
TC0100011527.hg.1 |
Clariom D TC |