Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005931.hg.1 |
TC07000445.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005931.hg.1 |
ENST00000427153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07005931.hg.1 |
NR_026690.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007988.hg.1 |
ENST00000427153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007988.hg.1 |
ENST00000427153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007988.hg.1 |
ENST00000427153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007988.hg.1 |
ENST00000427153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007988.hg.1 |
ENST00000427153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007988.hg.1 |
ENST00000427153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007988.hg.1 |
NR_026690.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007988.hg.1 |
NR_026690.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007988.hg.1 |
NR_026690.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007988.hg.1 |
NR_026690.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007988.hg.1 |
ENST00000427153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007988.hg.1 |
ENST00000427153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007988.hg.1 |
ENST00000427153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007988.hg.1 |
ENST00000427153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007988.hg.1 |
ENST00000427153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007988.hg.1 |
ENST00000427153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700007988.hg.1 |
NR_026690.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007988.hg.1 |
NR_026690.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007988.hg.1 |
NR_026690.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700007988.hg.1 |
NR_026690.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700155062.hg.1 |
ENST00000427153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700155062.hg.1 |
NR_026690.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700155062.hg.1 |
ENST00000427153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700155062.hg.1 |
NR_026690.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700155062.hg.1 |
TC0700007988.hg.1 |
Clariom D TC |