Assay Details
Gene Symbol: ABHD11-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026690.1 | 2 - 3 | 283 | 77 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BU595969.1 | - | 317 | Chr. - - [-] on Build | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | syntaxin 1A | STX1A |
HPC-1, P35-1, STX1, SYN1A |
-15.5kb | Anti-sense | Chr. 7 - 73699205 - 73719702 on NCBI Build GRCh38 |
Downstream | abhydrolase domain containing 11 | ABHD11 |
PP1226, WBSCR21 |
246.0 | Anti-sense | Chr. 7 - 73736094 - 73738867 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07000445.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3333480 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4721 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4722 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 50595 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3333480 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3333480 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05534228 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05534219 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25777851 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05534223 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05534230 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25777844 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05534220 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25777843 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05534227 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05534218 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25777847 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05534221 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05534227 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05534219 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25777851 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05534221 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05534223 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05534228 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25777844 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05534218 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05534220 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25777847 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25777843 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05534230 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534263 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534254 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534246 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534242 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15970423 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534271 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534231 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534265 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534253 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534262 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534259 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534217 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15970412 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534223 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534218 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15970425 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534251 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534267 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15970428 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15970421 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534244 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534270 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534227 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534226 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534248 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534219 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534215 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05534256 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15970413 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Genome U219 Array | 11757228_s_at | - | - |
Affymetrix Human Genome U219 Array | 11757228_PM_s_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR07005932.hg.1 | TC07000445.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07005932.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07005932.hg.1 | NR_026690.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007988.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007988.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007988.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007988.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007988.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007988.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700007988.hg.1 | NR_026690.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007988.hg.1 | NR_026690.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007988.hg.1 | NR_026690.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700007988.hg.1 | NR_026690.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07005933.hg.1 | TC07000445.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07005933.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07005933.hg.1 | NR_026690.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700155064.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700155064.hg.1 | NR_026690.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700155064.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700155064.hg.1 | NR_026690.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700155064.hg.1 | TC0700007988.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700007988.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007988.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007988.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007988.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007988.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007988.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700007988.hg.1 | NR_026690.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700007988.hg.1 | NR_026690.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700007988.hg.1 | NR_026690.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700007988.hg.1 | NR_026690.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700155065.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700155065.hg.1 | NR_026690.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700155065.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700155065.hg.1 | NR_026690.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700155065.hg.1 | TC0700007988.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700155063.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700155063.hg.1 | NR_026690.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700155063.hg.1 | ENST00000427153.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700155063.hg.1 | NR_026690.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700155063.hg.1 | TC0700007988.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A034537 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A034537 | - | - |
