Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08010398.hg.1 |
TC08000777.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08010398.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08010398.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08010400.hg.1 |
TC08000777.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08010400.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08010400.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08010400.hg.1 |
ENST00000524070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08010400.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000524070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000524070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000524070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
ENST00000524070.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009004.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800161419.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800161419.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800161419.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800161419.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800161419.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800161419.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800161419.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800161419.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800161419.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800161419.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800161419.hg.1 |
TC0800009004.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800161421.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800161421.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800161421.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800161421.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800161421.hg.1 |
ENST00000524070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800161421.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800161421.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800161421.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800161421.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800161421.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800161421.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800161421.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800161421.hg.1 |
ENST00000524070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800161421.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800161421.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800161421.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800161421.hg.1 |
TC0800009004.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000520101.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000522279.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000524070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000524070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000524070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
ENST00000524070.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_026706.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_120395.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_120396.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009004.hg.1 |
NR_120396.1 |
Refseq |