Assay Details
Gene Symbol: ZNF37BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026777.2 | 1 - 2 | 93 | 89 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 839 | LINC00839 | -57.1kb | Anti-sense | Chr. 10 - 42475491 - 42495337 on NCBI Build GRCh38 | |
Downstream | zinc finger protein 33B | ZNF33B |
KOX2, KOX31, ZNF11B |
36.2kb | Sense | Chr. 10 - 42574169 - 42638837 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001194.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10015003.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10015003.hg.1 | TC10001194.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10014997.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10014997.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10014997.hg.1 | TC10001194.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000452306.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000452306.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010408.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000170772.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000170772.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000170772.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000170772.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000170772.hg.1 | ENST00000435805.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000170772.hg.1 | ENST00000452075.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000170772.hg.1 | ENST00000473592.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000170772.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000170772.hg.1 | TC1000010408.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000170792.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000170792.hg.1 | NR_026777.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000170792.hg.1 | TC1000010408.hg.1 | Clariom D TC |
