Assay Details
Gene Symbol: LINC00661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026828.1 | 4 - 5 | 351 | 95 | ||
GenBank mRNA | CR749400.1 | - | 4 - 5 | 351 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000546512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000546512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000546512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000546512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000546512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000546512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000546512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000546512.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000547102.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000547102.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000547102.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000547102.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000547102.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000547102.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000547102.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000547102.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549131.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549131.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549131.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549131.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000552685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000552685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | ENST00000552685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000546512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000546512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000546512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000546512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000546512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000546512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000546512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000546512.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000547102.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000547102.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000547102.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000547102.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000547102.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000547102.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000547102.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000547102.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549131.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549131.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549131.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549131.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549354.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000549718.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000552608.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000552685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000552685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | ENST00000552685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | NR_026828.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007250.hg.1 | NR_026828.1 | Refseq |
