Assay Details
Gene Symbol: LOC151174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026926.1 | 2 - 3 | 459 | 103 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ILK associated serine/threonine phosphatase | ILKAP |
ILKAP2, ILKAP3, PP2C-DELTA, PPM1O |
-21.4kb | Sense | Chr. 2 - 238170401 - 238203702 on NCBI Build GRCh38 |
Downstream | TAR DNA binding protein pseudogene | LOC643387 | 3.9kb | Anti-sense | Chr. 2 - 238231686 - 238234345 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02002912.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000409070.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000409070.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000409070.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000409070.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000409070.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000409376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000409376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000409376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000409376.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000409942.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000409942.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000409942.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000409942.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000409942.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000466075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000466075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000466075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000466075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000466075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000466075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000466075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000466075.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000470346.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000470346.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000470346.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000470346.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000470346.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000470346.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000470346.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000470346.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000475669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000475669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000475669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000475669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000475669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | ENST00000475669.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | NR_026926.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | NR_026926.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | NR_026926.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | NR_026926.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | NR_026926.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | NR_026926.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | NR_026926.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016221.hg.1 | NR_026926.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02046734.hg.1 | TC02002912.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02046734.hg.1 | ENST00000334973.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02046734.hg.1 | ENST00000409070.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02046734.hg.1 | ENST00000409376.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02046734.hg.1 | ENST00000409942.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02046734.hg.1 | ENST00000470346.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02046734.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02046734.hg.1 | NR_026926.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200218973.hg.1 | ENST00000409070.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200218973.hg.1 | ENST00000409376.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200218973.hg.1 | ENST00000409942.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200218973.hg.1 | ENST00000470346.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200218973.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200218973.hg.1 | NR_026926.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200218973.hg.1 | ENST00000409070.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200218973.hg.1 | ENST00000409376.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200218973.hg.1 | ENST00000409942.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200218973.hg.1 | ENST00000470346.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200218973.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200218973.hg.1 | NR_026926.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200218973.hg.1 | TC0200016221.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000409070.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000409070.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000409070.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000409070.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000409070.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000409376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000409376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000409376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000409376.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000409942.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000409942.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000409942.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000409942.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000409942.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000466075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000466075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000466075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000466075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000466075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000466075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000466075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000466075.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000470346.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000470346.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000470346.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000470346.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000470346.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000470346.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000470346.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000470346.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000475669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000475669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000475669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000475669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000475669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | ENST00000475669.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | NR_026925.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | NR_026926.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | NR_026926.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | NR_026926.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | NR_026926.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | NR_026926.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | NR_026926.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | NR_026926.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016221.hg.1 | NR_026926.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 203780 | - | - |
