Assay Details
Gene Symbol: C20orf173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145350.1 | NP_001138822.1 | 1 - 2 | 94 | 75 | |
NR_026933.2 | 1 - 2 | 94 | 75 | |||
XM_006723710.2 | XP_006723773.1 | 1 - 2 | 94 | 75 | ||
XM_011528577.1 | XP_011526879.1 | 1 - 2 | 94 | 75 | ||
GenBank mRNA | AK310337.1 | - | 1 - 2 | 94 | 75 | |
DB057743.1 | - | 1 - 2 | 94 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001145350.1, XM_006723710.2, XM_011528577.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000424444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000424444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000424444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000246199.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000374345.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000422009.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000424444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000424444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000424444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000432650.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000444723.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | ENST00000450773.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NM_001145350.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008975.hg.1 | NR_026933.2 | Refseq |
