Assay Details
| Gene Symbol: PISRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027070.1 | 1 | 455 | 90 | ||
| GenBank mRNA | AW268472.1 | - | 1 | 455 | 90 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
blepharophimosis, epicanthus inversus and ptosis, candidate 1 (non-protein coding) |
BPESC1 | NCRNA00187 | -107.8kb | Anti-sense | Chr. 3 - 139104185 - 139125167 on NCBI Build GRCh38 |
| Downstream | mitochondrial ribosomal protein S22 | MRPS22 |
C3orf5, COXPD5, GIBT, GK002, MRP-S22, RPMS22 |
110.5kb | Anti-sense | Chr. 3 - 139343994 - 139357129 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001831.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4973 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4972 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759627 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759630 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759593 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759611 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759640 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618806 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618814 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15494075 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618803 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759618 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759581 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759609 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759602 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618798 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759587 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618794 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759577 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759633 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15494064 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759591 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759619 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759583 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618792 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15494073 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759628 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618784 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15494068 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759631 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618823 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759574 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759579 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618825 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759580 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618789 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759632 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759608 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759606 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759638 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15494081 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759629 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618822 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759612 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759628 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759627 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618784 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759602 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759579 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759640 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618826 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618806 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618814 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759574 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759629 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618799 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759577 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618822 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759587 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759593 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759609 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618829 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759591 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759619 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618818 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15494073 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618816 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759583 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15494075 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759618 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759612 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759638 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618789 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759631 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759581 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618794 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759630 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618803 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759632 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618792 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759608 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759633 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15494068 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618825 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618798 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759580 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759611 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22759606 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15494064 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618823 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618834 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15494081 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02618787 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618780 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618838 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618826 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618741 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618762 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15494067 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15494064 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15494057 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618726 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618753 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618737 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618805 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15494071 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618766 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15494050 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618773 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618778 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15494075 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618790 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618770 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618764 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618754 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618833 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618751 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618730 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618760 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618818 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618801 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618829 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15494073 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15494053 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618831 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618812 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15494054 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15494069 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618758 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618739 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618823 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618803 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15494063 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15494066 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618821 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618806 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618756 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618786 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15494051 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618746 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618749 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618728 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618808 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618839 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618777 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618835 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618815 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618735 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618782 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618799 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02618798 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Genome U219 Array | 11757257_at | - | - |
| Affymetrix Human Genome U219 Array | 11757257_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR03033119.hg.1 | TC03001831.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03033119.hg.1 | NR_027070.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008967.hg.1 | ENST00000616930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008967.hg.1 | ENST00000626175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008967.hg.1 | ENST00000626175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194161.hg.1 | NR_027070.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300194161.hg.1 | NR_027070.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300194161.hg.1 | TC0300012593.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008967.hg.1 | ENST00000616930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008967.hg.1 | ENST00000626175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008967.hg.1 | ENST00000626175.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021665 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021665 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3309709 | NR_027070.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3309709 | AI204197.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3309709 | AW268472.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309709 | NR_027070.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309709 | AI204197.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3309709 | AW268472.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309709 | NR_027070.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309709 | AI204197.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3309709 | AW268472.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309709 | NR_027070.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309709 | AI204197.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3309709 | AW268472.1 | human_transcriptome |

PISRT1
Genomic Map