Assay Details
| Gene Symbol: LINC00486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027099.1 | 1 - 2 | 120 | 93 | ||
| GenBank mRNA | BC040883.2 | - | 2 - 3 | 340 | 93 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BM548532.1 | - | 371 | Chr. - - [-] on Build | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tetratricopeptide repeat domain 27 | TTC27 | -61.6kb | Sense | Chr. 2 - 32628020 - 32821051 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100271832 | LOC100271832 | 44.1kb | Sense | Chr. 2 - 32927127 - 32946136 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000217.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02003212.hg.1 | TC02000217.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003212.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007199.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003204.hg.1 | TC02000217.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003204.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003204.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003200.hg.1 | TC02000217.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003200.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003200.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003202.hg.1 | TC02000217.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003202.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003202.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003202.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003202.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003203.hg.1 | TC02000217.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003203.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003203.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149712.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149712.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149712.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149712.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149712.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149712.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149712.hg.1 | TC0200007199.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200149713.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149713.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149713.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149713.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149713.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149713.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149713.hg.1 | TC0200007199.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000414054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | ENST00000440786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027097.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027098.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007199.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149731.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149731.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149731.hg.1 | TC0200007199.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200149733.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149733.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149733.hg.1 | TC0200007199.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200149710.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149710.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149710.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149710.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149710.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149710.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149710.hg.1 | TC0200007199.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200149711.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149711.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149711.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149711.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149711.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149711.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149711.hg.1 | ENST00000424364.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149711.hg.1 | ENST00000435075.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200149711.hg.1 | NR_027099.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149711.hg.1 | NR_027100.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200149711.hg.1 | TC0200007199.hg.1 | Clariom D TC |

LINC00486
Genomic Map