Assay Details
Gene Symbol: LOC285692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027112.2 | 7 - 8 | 968 | 99 | ||
GenBank mRNA | BC031253.1 | - | 7 - 8 | 968 | 99 | |
BC047112.1 | - | 7 - 8 | 968 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | taste 2 receptor member 1 | TAS2R1 |
T2R1, TRB7 |
-82.7kb | Sense | Chr. 5 - 9628997 - 9630351 on NCBI Build GRCh38 |
Downstream | family with sequence similarity 173 member B | FAM173B | JS-2 | 511.7kb | Sense | Chr. 5 - 10225507 - 10249909 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05003421.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05016536.hg.1 | TC05003421.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05016536.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05016536.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05016538.hg.1 | TC05003421.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05016538.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05016538.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000606169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000606169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000606169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000606169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000606222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000606222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000606222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000606222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000606222.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000606744.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000606744.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | ENST00000606744.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500195254.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500195254.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500195254.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500195254.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500195254.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500195254.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500195254.hg.1 | TC0500013289.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000512976.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000606169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000606169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000606169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000606169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000606222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000606222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000606222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000606222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000606222.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000606744.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000606744.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | ENST00000606744.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013289.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500195253.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500195253.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500195253.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500195253.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500195253.hg.1 | ENST00000511616.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500195253.hg.1 | NR_027112.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500195253.hg.1 | TC0500013289.hg.1 | Clariom D TC |
