Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006501.hg.1 |
ENST00000314040.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006501.hg.1 |
ENST00000314040.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600006501.hg.1 |
ENST00000314040.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014419.hg.1 |
TC06001211.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014419.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014419.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014420.hg.1 |
TC06001211.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014420.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06014420.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200017.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200017.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200017.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200017.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200017.hg.1 |
TC0600014212.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600200015.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200015.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200015.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200015.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200015.hg.1 |
TC0600014212.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600006501.hg.1 |
ENST00000314040.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006501.hg.1 |
ENST00000314040.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600006501.hg.1 |
ENST00000314040.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600200006.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200006.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200006.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200006.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200006.hg.1 |
TC0600014212.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600200014.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200014.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200014.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200014.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200014.hg.1 |
TC0600014212.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600200016.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200016.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200016.hg.1 |
NR_027115.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200016.hg.1 |
NR_027116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600200016.hg.1 |
TC0600014212.hg.1 |
Clariom D TC |