Assay Details
Gene Symbol: LINC01622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027115.3 | 2 - 3 | 1755 | 92 | ||
GenBank mRNA | BC117448.1 | - | 2 - 3 | 1418 | 92 | |
BC126314.1 | - | 2 - 3 | 1418 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC101927353 | LOC101927353 | -82.8kb | Anti-sense | Chr. 6 - 906480 - 915608 on NCBI Build GRCh38 | |
Downstream | uncharacterized LOC107986554 | LOC107986554 | 300.5kb | Sense | Chr. 6 - 1306011 - 1312404 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001211.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
