Assay Details
Gene Symbol: LOC728743 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027237.1 | 1 - 2 | 155 | 97 | ||
GenBank mRNA | AI862583.1 | - | 1 - 2 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger protein 775 | ZNF775 | -7.1kb | Sense | Chr. 7 - 150379317 - 150398631 on NCBI Build GRCh38 | |
Downstream | long intergenic non-protein coding RNA 996 | LINC00996 | 25.2kb | Sense | Chr. 7 - 150433654 - 150448140 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07003296.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000464630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000464630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000464630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000464630.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000480389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000480389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000480389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07015226.hg.1 | TC07003296.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07015226.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07015227.hg.1 | TC07003296.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07015227.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07015228.hg.1 | TC07003296.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07015228.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000464630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000464630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000464630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000464630.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000476489.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000480389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000480389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000480389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013490.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700199501.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199501.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199501.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199501.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700199501.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199501.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199501.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199501.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700199501.hg.1 | TC0700013490.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700199502.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199502.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199502.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199502.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700199502.hg.1 | ENST00000481877.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199502.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199502.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199502.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700199502.hg.1 | TC0700013490.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700199503.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199503.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199503.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700199503.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199503.hg.1 | ENST00000498682.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199503.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700199503.hg.1 | TC0700013490.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700199497.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199497.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199497.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700199497.hg.1 | ENST00000474836.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199497.hg.1 | ENST00000491664.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700199497.hg.1 | NR_027237.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700199497.hg.1 | TC0700013490.hg.1 | Clariom D TC |
