Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010941.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010941.hg.1 |
TC10000893.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010939.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010939.hg.1 |
TC10000893.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000425266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000425266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
ENST00000425266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162870.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162870.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162870.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162870.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162870.hg.1 |
TC1000009163.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000162875.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162875.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162875.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162875.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162875.hg.1 |
TC1000009163.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000162869.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162869.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162869.hg.1 |
TC1000009163.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000368895.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000425266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000425266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
ENST00000425266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009163.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162871.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162871.hg.1 |
NR_027282.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162871.hg.1 |
TC1000009163.hg.1 |
Clariom D TC |