Assay Details
| Gene Symbol: FAM201A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027294.1 | 1 - 2 | 279 | 103 | ||
| GenBank mRNA | BC036230.1 | - | 1 - 2 | 279 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ankyrin repeat domain 18A | ANKRD18A | -433.0 | Anti-sense | Chr. 9 - 38566260 - 38620587 on NCBI Build GRCh38 | |
| Downstream | contactin associated protein-like 3 | CNTNAP3 |
CASPR3, CNTNAP3A |
446.9kb | Anti-sense | Chr. 9 - 39072767 - 39288303 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000225.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27002513 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27002509 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27002517 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27002514 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27002487 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06712041 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16177526 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27002498 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06712033 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06712036 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16177521 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27002519 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06712045 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06712035 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06712040 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27002515 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P27002518 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06712032 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16177534 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P06712037 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 391662 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 348818 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3381328 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P886197 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 8464 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P125308 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P125308 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 35280 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3381328 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3381328 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16177534 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16177526 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06712035 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16177521 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27002487 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06712034 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27002498 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06712032 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06712037 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06712035 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16177526 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06712034 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27002487 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27002498 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06712032 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06712037 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16177534 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16177521 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P886197 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P125308 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P125308 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P125308 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P886197 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 5421 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 16613 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P38715158 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P233629 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P834920 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16177521 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06712046 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06712040 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06712033 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06712032 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16177523 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06712044 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06712042 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16177518 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P06712035 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16177504 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16177507 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16177526 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P38715158 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1557541_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1557014_PM_a_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002628.hg.1 | TC09000225.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002628.hg.1 | ENST00000377680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002628.hg.1 | ENST00000484285.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002628.hg.1 | NR_027294.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007195.hg.1 | ENST00000377680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007195.hg.1 | ENST00000377680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007195.hg.1 | ENST00000377680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007195.hg.1 | ENST00000377680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007195.hg.1 | ENST00000471864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007195.hg.1 | ENST00000471864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007195.hg.1 | ENST00000484285.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007195.hg.1 | ENST00000484285.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007195.hg.1 | ENST00000484285.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007195.hg.1 | NR_027294.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007195.hg.1 | NR_027294.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007195.hg.1 | NR_027294.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002629.hg.1 | TC09000225.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002629.hg.1 | ENST00000377680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002629.hg.1 | ENST00000484285.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002629.hg.1 | NR_027294.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002630.hg.1 | TC09000225.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002630.hg.1 | ENST00000377680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002630.hg.1 | ENST00000484285.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002630.hg.1 | NR_027294.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002627.hg.1 | TC09000225.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002627.hg.1 | ENST00000377680.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09002627.hg.1 | NR_027294.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900149023.hg.1 | ENST00000377680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900149023.hg.1 | ENST00000471864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900149023.hg.1 | ENST00000484285.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900149023.hg.1 | NR_027294.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900149023.hg.1 | ENST00000377680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900149023.hg.1 | ENST00000471864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900149023.hg.1 | ENST00000484285.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900149023.hg.1 | NR_027294.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900149023.hg.1 | TC0900007195.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900007195.hg.1 | ENST00000377680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007195.hg.1 | ENST00000377680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007195.hg.1 | ENST00000377680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007195.hg.1 | ENST00000377680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007195.hg.1 | ENST00000471864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007195.hg.1 | ENST00000471864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007195.hg.1 | ENST00000484285.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007195.hg.1 | ENST00000484285.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007195.hg.1 | ENST00000484285.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007195.hg.1 | NR_027294.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007195.hg.1 | NR_027294.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007195.hg.1 | NR_027294.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900149024.hg.1 | ENST00000377680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900149024.hg.1 | ENST00000484285.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900149024.hg.1 | NR_027294.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900149024.hg.1 | ENST00000377680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900149024.hg.1 | ENST00000484285.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900149024.hg.1 | NR_027294.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900149024.hg.1 | TC0900007195.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900149022.hg.1 | ENST00000377680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900149022.hg.1 | NR_027294.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900149022.hg.1 | ENST00000377680.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900149022.hg.1 | NR_027294.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900149022.hg.1 | TC0900007195.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1557541_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1557014_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1557014_PM_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1557541_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557014_a_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557014_a_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557541_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557014_a_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557014_a_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557541_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557541_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557014_a_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557014_a_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557541_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557541_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557014_a_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557014_a_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557541_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557541_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557541_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557541_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557541_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557014_a_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557014_a_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557541_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1557014_a_at3 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A038585 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A038585 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 446972 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1744581 | BC036230.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1744581 | BC036230.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1744581 | BC036230.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1744581 | BC036230.1 | human_transcriptome |

FAM201A
Genomic Map