Assay Details
Gene Symbol: LINC00261 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_001558.3 | 2 - 3 | 223 | 93 | ||
GenBank mRNA | BG655894.1 | - | 2 - 3 | 227 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC284788 | LOC284788 | -146.0kb | Sense | Chr. 20 - 22400333 - 22420643 on NCBI Build GRCh38 | |
Downstream | forkhead box A2 | FOXA2 |
HNF3B, TCF3B |
13.1kb | Sense | Chr. 20 - 22581004 - 22585463 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000697.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR20009599.hg.1 | ENST00000450346.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20009599.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20009599.hg.1 | TC20000697.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20009601.hg.1 | ENST00000450346.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20009601.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20009601.hg.1 | TC20000697.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20009600.hg.1 | ENST00000450346.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20009600.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20009600.hg.1 | TC20000697.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000420070.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000420070.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000420070.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000420070.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000609300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000609300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000609300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | ENST00000609300.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000160479.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000160479.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000160479.hg.1 | TC2000008635.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000160479.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000160479.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000160476.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000160476.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000160476.hg.1 | TC2000008635.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000160476.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000160476.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000160480.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000160480.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000160480.hg.1 | TC2000008635.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000160480.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000160480.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000420070.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000420070.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000420070.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000420070.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000609300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000609300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000609300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | ENST00000609300.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008635.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000160478.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000160478.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000160478.hg.1 | TC2000008635.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000160478.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000160478.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000160470.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000160470.hg.1 | NR_001558.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000160470.hg.1 | TC2000008635.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000160470.hg.1 | ENST00000564492.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000160470.hg.1 | NR_001558.3 | Refseq |
