Assay Details
Gene Symbol: LINC00893 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027455.5 | 8 - 9 | 1518 | 96 | ||
GenBank mRNA | AK124040.1 | - | 8 - 9 | 1518 | 96 | |
U66044.1 | - | 2 - 3 | 279 | 96 | ||
U66045.1 | - | 2 - 3 | 279 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized IDS2 | IDS2 | -2.8kb | Sense | Chr. X - 149511554 - 149526523 on NCBI Build GRCh38 | |
Downstream | chromosome X open reading frame 40A | CXorf40A |
CXorf40, EOLA1, FLJ16423 |
11.1kb | Anti-sense | Chr. X - 149540630 - 149555338 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
