Assay Details
| Gene Symbol: SNORD37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002602.1 | 1 | 29 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | eukaryotic translation elongation factor 2 | EEF2 |
EEF-2, EF-2, EF2, SCA26 |
Sense | Chr. 19 - 3976056 - 3985463 on NCBI Build GRCh38 | |
| Downstream | protein inhibitor of activated STAT 4 | PIAS4 |
PIAS-gamma, PIASY, Piasg, ZMIZ6 |
25.0kb | Anti-sense | Chr. 19 - 4007598 - 4039386 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000384048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000384048.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000594885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000594885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000594885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000594885.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000596417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000596417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000596417.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000600720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000600720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000600720.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000600794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000600794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000600794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | ENST00000600794.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NR_002602.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009316.hg.1 | NR_002602.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000309311.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000384048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000384048.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000594885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000594885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000594885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000594885.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000596417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000596417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000596417.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000598436.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000600720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000600720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000600720.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000600794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000600794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000600794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | ENST00000600794.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NM_001961.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NR_002602.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009316.hg.1 | NR_002602.1 | Refseq |

SNORD37
Genomic Map