Assay Details
Gene Symbol: LINC00905 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_024335.2 | 3 - 4 | 896 | 86 | ||
NR_110321.1 | 3 - 4 | 606 | 86 | |||
GenBank mRNA | AK058057.1 | - | 3 - 4 | 855 | 86 | |
BC031284.1 | - | 3 - 4 | 606 | 86 | ||
BC069223.1 | - | 3 - 4 | 828 | 86 | ||
BC104221.2 | - | 2 - 3 | 793 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 661 | LINC00661 | -8.0kb | Sense | Chr. 19 - 16015634 - 16027462 on NCBI Build GRCh38 | |
Downstream | tropomyosin 4 | TPM4 | HEL-S-108 | 31.5kb | Sense | Chr. 19 - 16067507 - 16103005 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
