Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003974.hg.1 |
ENST00000397365.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003974.hg.1 |
NR_024335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003974.hg.1 |
TC19000281.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900152432.hg.1 |
ENST00000588117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152432.hg.1 |
ENST00000588117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152432.hg.1 |
ENST00000588838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152432.hg.1 |
ENST00000588838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152432.hg.1 |
NR_024335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900152432.hg.1 |
NR_110321.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900152432.hg.1 |
TC1900007249.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900152432.hg.1 |
ENST00000588117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152432.hg.1 |
ENST00000588117.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152432.hg.1 |
ENST00000588838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152432.hg.1 |
ENST00000588838.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152432.hg.1 |
NR_024335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900152432.hg.1 |
NR_110321.1 |
Refseq |