Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008003.hg.1 |
ENST00000580491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008003.hg.1 |
ENST00000580491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008003.hg.1 |
ENST00000580491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008003.hg.1 |
ENST00000580491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008003.hg.1 |
NR_024419.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008003.hg.1 |
NR_024419.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008003.hg.1 |
NR_024419.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008003.hg.1 |
NR_024419.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003683.hg.1 |
NR_024419.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003683.hg.1 |
TC18000314.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003682.hg.1 |
NR_024419.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18003682.hg.1 |
TC18000314.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800154751.hg.1 |
ENST00000580491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800154751.hg.1 |
NR_024419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800154751.hg.1 |
TC1800008003.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800154751.hg.1 |
ENST00000580491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800154751.hg.1 |
NR_024419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008003.hg.1 |
ENST00000580491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008003.hg.1 |
ENST00000580491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008003.hg.1 |
ENST00000580491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008003.hg.1 |
ENST00000580491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008003.hg.1 |
NR_024419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008003.hg.1 |
NR_024419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008003.hg.1 |
NR_024419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008003.hg.1 |
NR_024419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800154752.hg.1 |
ENST00000580491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800154752.hg.1 |
NR_024419.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800154752.hg.1 |
TC1800008003.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800154752.hg.1 |
ENST00000580491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800154752.hg.1 |
NR_024419.1 |
Refseq |