Assay Details
Gene Symbol: BCRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_024494.1 | 1 - 2 | 530 | 76 | ||
GenBank mRNA | AK297038.1 | - | 1 - 2 | 530 | 76 | |
BM920079.1 | - | 1 - 2 | 507 | 76 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | gamma-glutamyltransferase 1 | GGT1 |
CD224, D22S672, D22S732, GGT, GGT 1, GTG |
-3.9kb | Sense | Chr. 22 - 24583750 - 24629005 on NCBI Build GRCh38 |
Downstream |
POM121 transmembrane nucleoporin like 10, pseudogene |
POM121L10P | 367.0 | Anti-sense | Chr. 22 - 24645166 - 24659147 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000153.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK297038.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 440514 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 663676 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 733968 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 656310 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 623714 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 11688 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 264310 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 247599 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 196321 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 192268 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 268794 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P873764 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P127235 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447247 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11447247 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447248 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P11447247 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8074732 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8074731 | - | - |
Affymetrix Human Genome U219 Array | 11761044_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11761044_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR22002953.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22002953.hg.1 | TC22000153.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22002952.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22002952.hg.1 | TC22000153.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000611425.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000611425.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000611425.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000611425.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000611425.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000614808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000614808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000614808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200006900.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22002958.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22002958.hg.1 | TC22000153.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2200007930.hg.1 | ENST00000621762.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007930.hg.1 | ENST00000621762.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22002954.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22002954.hg.1 | TC22000153.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22002955.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22002955.hg.1 | TC22000153.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2000008689.hg.1 | ENST00000447582.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22002956.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22002956.hg.1 | TC22000153.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22002957.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22002957.hg.1 | TC22000153.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2200147894.hg.1 | TC2200006900.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200147894.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147894.hg.1 | ENST00000611425.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147894.hg.1 | ENST00000614808.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147894.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147894.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147894.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147894.hg.1 | ENST00000611425.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147894.hg.1 | ENST00000614808.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147894.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147894.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008689.hg.1 | ENST00000447582.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000611425.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000611425.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000611425.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000611425.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000611425.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000614808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000614808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000614808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200006900.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147880.hg.1 | TC2200006900.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200147880.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147880.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147880.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147880.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147880.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147880.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147890.hg.1 | TC2200006900.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200147890.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147890.hg.1 | ENST00000611425.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147890.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147890.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147890.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147890.hg.1 | ENST00000611425.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147890.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147890.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007930.hg.1 | ENST00000621762.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007930.hg.1 | ENST00000621762.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147879.hg.1 | TC2200006900.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200147879.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147879.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147879.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147879.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147883.hg.1 | TC2200006900.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200147883.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147883.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147883.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147883.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147883.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147883.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147883.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147883.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147884.hg.1 | TC2200006900.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200147884.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147884.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147884.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147884.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147884.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147884.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147884.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147884.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147899.hg.1 | TC2200006900.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200147899.hg.1 | ENST00000614808.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147899.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147899.hg.1 | ENST00000614808.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147899.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147882.hg.1 | TC2200006900.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200147882.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147882.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147882.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147882.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147882.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147882.hg.1 | ENST00000447261.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147882.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147882.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147891.hg.1 | TC2200006900.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200147891.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147891.hg.1 | ENST00000611425.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147891.hg.1 | ENST00000614808.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147891.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147891.hg.1 | NR_024494.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200147891.hg.1 | ENST00000382243.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147891.hg.1 | ENST00000611425.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147891.hg.1 | ENST00000614808.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147891.hg.1 | ENST00000621462.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200147891.hg.1 | NR_024494.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 226602_PM_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 1553591_at | - | - |
Human Genome U133 Plus 2.0 Array | 1553592_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 226602_s_at | - | - |
Human Genome U133 Plus 2.0 Array | 1553591_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1553592_PM_x_at | - | - |
Human Genome U95 Set | 64904_PM_at | - | - |
Human Genome U95 Set | 64904_at | - | - |
Human Genome U95 Set | 91641_g_at | - | - |
Human Genome U95 Set | 91639_PM_at | - | - |
Human Genome U95 Set | 91641_PM_g_at | - | - |
Human Genome U95 Set | 91639_at | - | - |
Human Genome U133 Set | 226602_s_at | - | - |
Human Genome U133 Set | 226602_PM_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020237 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A020237 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1792592 | NR_024593.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1792592 | NR_024593.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1792592 | NR_024593.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1792592 | NR_024593.1 | human_transcriptome |
