Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
ENST00000419779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
ENST00000419779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
ENST00000419779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
ENST00000419779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
ENST00000423162.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
ENST00000423162.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
ENST00000423162.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
ENST00000423162.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
ENST00000423162.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
ENST00000423162.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
ENST00000423162.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012446.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002954.hg.1 |
ENST00000423162.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002954.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10002954.hg.1 |
TC10000254.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
ENST00000624635.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
ENST00000472090.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
ENST00000624635.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010407.hg.1 |
NR_103829.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
ENST00000419779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
ENST00000419779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
ENST00000419779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
ENST00000419779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
ENST00000423162.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
ENST00000423162.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
ENST00000423162.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
ENST00000423162.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
ENST00000423162.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
ENST00000423162.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
ENST00000423162.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012446.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185440.hg.1 |
ENST00000423162.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185440.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185440.hg.1 |
ENST00000423162.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000185440.hg.1 |
NR_024524.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000185440.hg.1 |
TC1000012446.hg.1 |
Clariom D TC |