Assay Details
| Gene Symbol: ARL6IP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024526.2 | 2 - 3 | 148 | 120 | ||
| GenBank mRNA | AW967583.1 | - | 2 - 3 | 103 | 120 | |
| BP200063.1 | - | 2 - 3 | 132 | 120 | ||
| BX437990.2 | - | 2 - 3 | 149 | 120 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | pre-mRNA processing factor 40 homolog A | PRPF40A |
FBP-11, FBP11, FLAF1, FNBP3, HIP-10, HIP10, HYPA, NY-REN-6, Prp40 |
-2.6kb | Anti-sense | Chr. 2 - 152651593 - 152718007 on NCBI Build GRCh38 |
| Downstream |
reprimo, TP53 dependent G2 arrest mediator candidate |
RPRM | REPRIMO | 742.2kb | Anti-sense | Chr. 2 - 153477338 - 153478808 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000942.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02013802.hg.1 | TC02000942.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013802.hg.1 | ENST00000326446.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013802.hg.1 | ENST00000463690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013802.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02013802.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000425034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000425034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000425034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000425034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000425034.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000455875.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000455875.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000455875.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000455875.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000455875.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000455875.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000455875.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000463690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000463690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000463690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000463690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000463690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000463690.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000495469.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000495469.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000495469.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000495469.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000495469.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000495469.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | ENST00000495469.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009687.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167697.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167697.hg.1 | ENST00000425034.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167697.hg.1 | ENST00000455875.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167697.hg.1 | ENST00000463690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167697.hg.1 | ENST00000495469.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167697.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167697.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167697.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167697.hg.1 | ENST00000425034.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167697.hg.1 | ENST00000455875.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167697.hg.1 | ENST00000463690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167697.hg.1 | ENST00000495469.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200167697.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167697.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200167697.hg.1 | TC0200009687.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000326446.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000425034.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000425034.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000425034.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000425034.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000425034.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000455875.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000455875.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000455875.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000455875.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000455875.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000455875.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000455875.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000463690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000463690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000463690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000463690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000463690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000463690.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000495469.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000495469.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000495469.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000495469.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000495469.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000495469.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | ENST00000495469.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NM_152522.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NR_024526.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009687.hg.1 | NR_024526.2 | Refseq |

ARL6IP6
Genomic Map