Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013132.hg.1 |
TC01000825.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013132.hg.1 |
ENST00000466454.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01013132.hg.1 |
NR_024604.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163694.hg.1 |
ENST00000284054.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163694.hg.1 |
ENST00000466454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163694.hg.1 |
ENST00000490028.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163694.hg.1 |
NR_024604.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163694.hg.1 |
ENST00000284054.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163694.hg.1 |
ENST00000466454.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163694.hg.1 |
ENST00000490028.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100163694.hg.1 |
NR_024604.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100163694.hg.1 |
TC0100008925.hg.1 |
Clariom D TC |