Assay Details
Gene Symbol: LOC494141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026541.1 | 1 - 2 | 58 | 73 | ||
NR_026564.1 | 1 - 2 | 58 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MAS related GPR family member X4 | MRGPRX4 |
GPCR, MRGX4, SNSR6 |
-34.9kb | Sense | Chr. 11 - 18172837 - 18174280 on NCBI Build GRCh38 |
Downstream |
serum amyloid A4, constitutive |
SAA4 |
C-SAA, CSAA |
19.8kb | Anti-sense | Chr. 11 - 18231355 - 18236837 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000237.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11003113.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11003113.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11003113.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003113.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003113.hg.1 | TC11000237.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11003116.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11003116.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11003116.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11003116.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003116.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003116.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003116.hg.1 | TC11000237.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11003117.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11003117.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003117.hg.1 | TC11000237.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000623626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000623626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149206.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149206.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149206.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149206.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149206.hg.1 | TC1100006992.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100149200.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149200.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149200.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149200.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149200.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149200.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149200.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149200.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149200.hg.1 | TC1100006992.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100149203.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149203.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149203.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149203.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149203.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149203.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149203.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149203.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149203.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149203.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149203.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149203.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149203.hg.1 | TC1100006992.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100149204.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149204.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149204.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149204.hg.1 | ENST00000623626.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149204.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149204.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149204.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149204.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149204.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149204.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149204.hg.1 | ENST00000623626.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149204.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149204.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149204.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149204.hg.1 | TC1100006992.hg.1 | Clariom D TC |
