Assay Details
Gene Symbol: LOC494141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026563.1 | 1 - 2 | 1384 | 91 | ||
NR_026564.1 | 2 - 3 | 1239 | 91 | |||
GenBank mRNA | AK131059.1 | - | 2 - 3 | 1217 | 91 | |
BC026079.1 | - | 1 - 2 | 1391 | 91 | ||
BC034967.1 | - | 1 - 2 | 1391 | 91 | ||
BC044592.1 | - | 1 - 2 | 1384 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | MAS related GPR family member X4 | MRGPRX4 |
GPCR, MRGX4, SNSR6 |
-35.0kb | Sense | Chr. 11 - 18172837 - 18174280 on NCBI Build GRCh38 |
Downstream |
serum amyloid A4, constitutive |
SAA4 |
C-SAA, CSAA |
17.8kb | Anti-sense | Chr. 11 - 18231355 - 18236837 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000237.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28071081 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28071049 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07671190 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07671192 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16424904 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28071065 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28071050 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07671188 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16424903 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28071055 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P07671187 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28071079 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 99246 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 723393 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 82727 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 230726 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 391752 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4666 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4665 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P02402912 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P112512 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28071060 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28071055 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28071065 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671188 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16424889 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16424890 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28071052 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671187 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28071054 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28071050 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28071052 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28071060 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28071050 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28071054 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16424889 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671187 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07671188 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16424890 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28071055 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28071065 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P177270 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671191 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671183 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671190 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16424873 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671193 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671188 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671187 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671181 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16424875 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671189 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671192 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P07671195 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P11013245 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7938753 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7938751 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7938755 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7938754 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7938752 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7938750 | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR11003118.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11003118.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11003118.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003118.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003118.hg.1 | TC11000237.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | ENST00000623626.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003115.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11003115.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003115.hg.1 | TC11000237.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100149202.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149202.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149202.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149202.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149202.hg.1 | TC1100006992.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | ENST00000623626.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006992.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149205.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149205.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149205.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149205.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149205.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149205.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149205.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149205.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149205.hg.1 | ENST00000534640.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149205.hg.1 | NR_026541.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149205.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149205.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149205.hg.1 | TC1100006992.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100149208.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149208.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149208.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149208.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149208.hg.1 | ENST00000340135.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149208.hg.1 | ENST00000527059.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149208.hg.1 | NR_026563.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149208.hg.1 | NR_026564.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149208.hg.1 | TC1100006992.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046191 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046192 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046190 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046190 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046192 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046191 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3292784 | BC026082.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3292784 | NR_026541.1 | human_transcriptome |
