Assay Details
| Gene Symbol: RPL13AP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026715.1 | 1 | 553 | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | BBSome interacting protein 1 | BBIP1 |
BBIP10, BBS18, NCRNA00081, bA348N5.3 |
-17.2kb | Sense | Chr. 10 - 110898730 - 110919366 on NCBI Build GRCh38 |
| Downstream | microRNA 548e | MIR548E |
MIRN548E, hsa-mir-548e |
51.7kb | Anti-sense | Chr. 10 - 110988926 - 110989013 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001661.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3340060 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P272061 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869598 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869576 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869593 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07478862 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869587 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869575 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869604 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869614 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869597 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869580 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07478825 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07478868 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869585 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869573 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869578 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869569 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869605 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07478864 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869595 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869591 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869581 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869577 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869601 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869570 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07478866 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869600 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869584 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869599 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869585 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869576 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869598 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869578 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869584 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869587 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869614 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07478825 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869595 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869569 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869575 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869580 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869599 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869591 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869604 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869573 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869597 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869570 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869600 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869593 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07478862 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869577 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07478868 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869605 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07478866 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869601 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07478864 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27869581 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478780 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478839 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478832 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478830 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478863 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478858 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478837 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478831 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478851 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478773 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478861 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478866 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478868 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478782 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478776 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478827 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478848 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478842 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478840 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478870 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478854 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478774 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478856 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478836 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478779 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07478844 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7936321 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7936320 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021786.hg.1 | NR_026715.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021786.hg.1 | TC10001661.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000181008.hg.1 | ENST00000430133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181008.hg.1 | NR_026715.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181008.hg.1 | ENST00000430133.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000181008.hg.1 | NR_026715.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000181008.hg.1 | TC1000011827.hg.1 | Clariom D TC |
| Human Genome U95 Set | 74560_PM_f_at | - | - |
| Human Genome U95 Set | 74560_f_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044342 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A043301 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A043301 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A044342 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 157784 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 234426 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1789809 | NR_026715.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1789809 | NR_026715.1 | human_transcriptome |

RPL13AP6
Genomic Map