Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000489670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000489670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000489670.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300159062.hg.1 |
ENST00000497996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300159062.hg.1 |
NR_109841.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300159062.hg.1 |
ENST00000497996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300159062.hg.1 |
NR_109841.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300159062.hg.1 |
TC0300008303.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000489670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000489670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000489670.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
ENST00000497996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109841.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300008303.hg.1 |
NR_109842.1 |
Refseq |